Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR3/TNFRSF25 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen DR3/TNFRSF25
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-2 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056380T
SDP
View Documents
Novus Biologicals
NB10056380SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056380
SDP
View Documents
Novus Biologicals
NB10056380
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DR3/TNFRSF25
Western Blot 0.5-2 ug/ml
Polyclonal
Purified
RUO
Human
NP_042997
8718
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 398-417 of human DR3 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Apoptosis, Cell Biology
PBS containing 0.2% gelatin with 0.05% Sodium Azide
Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1
TNFRSF25
IgG
Protein G purified
In HeLa, a 50 kDa band should be observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.