Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR6/TNFRSF21 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB057474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057474 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057474 Supplier Novus Biologicals Supplier No. NBP299035FR

Rabbit Polyclonal Antibody

DR6/TNFRSF21 Polyclonal specifically detects DR6/TNFRSF21 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen DR6/TNFRSF21
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant DR6/TNFRSF21 (NP_055267.1; Met1-Leu350).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 27242
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.