Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR6/TNFRSF21 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB321236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB321236 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB321236 Supplier Novus Biologicals Supplier No. NBP298249PE

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

DR6/TNFRSF21 Polyclonal antibody specifically detects DR6/TNFRSF21 in Mouse samples. It is validated for ELISA

Specifications

Antigen DR6/TNFRSF21
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse DR6/TNFRSF21 (Accession#: NP_848704.1; Met 1-His 349)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 27242
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.