Learn More
Dräger™ Short-Term Detector Tubes: Aniline
Detector tubes
Specifications
Description Detector Tubes
Components Measured Aniline
Quantity 10
View More Specs
Products 2
Catalog Number Mfr. No. Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
Catalog Number Mfr. No. Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
17-985-180
View Documents
Dräger™
6733171
Aniline Aniline 0.5/a 0.5 to 10 7 to 12mg/L N/A N/A N/A
17-985-181
View Documents
Dräger™
CH20401
Aniline Aniline 5/a 1 to 20 Less than50mg/L N/A N/A N/A
Description
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Detector Tubes
Aniline
10
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.