Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRAM Antibody (M3-P4B4), HRP, Novus Biologicals™
SDP

Catalog No. NBP250379H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250379H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250379H Supplier Novus Biologicals Supplier No. NBP250379H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DRAM Monoclonal specifically detects DRAM in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen DRAM
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M3-P4B4
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias Damage-regulated autophagy modulator, DNA damage-regulated autophagy modulator protein 1, DNA-damage regulated autophagy modulator 1, DRAMFLJ11259
Gene Symbols DRAM1
Host Species Mouse
Immunogen Peptide Sequence - EKDYVYHVVS (amino acids 192 - 201)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 55332
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.