Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRAM Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB357466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357466 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357466 Supplier Novus Biologicals Supplier No. NBP176759PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DRAM Polyclonal antibody specifically detects DRAM in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Antigen DRAM
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation PBS
Gene Alias Damage-regulated autophagy modulator, DNA damage-regulated autophagy modulator protein 1, DNA-damage regulated autophagy modulator 1, DRAMFLJ11259
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the amino terminus of human DRAM. The immunogen is located within amino acids 30-80 of DRAM. Amino Acid Sequence: LPYISDTGTTPPESGI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 55332
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.