Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Draxin/C1orf187 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB042025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042025 Supplier Novus Biologicals Supplier No. NBP297115AF532

Rabbit Polyclonal Antibody

Draxin/C1orf187 Polyclonal specifically detects Draxin/C1orf187 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Draxin/C1orf187
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias AGPA3119, C1orf187, chromosome 1 open reading frame 187, dorsal inhibitory axon guidance protein, Dorsal repulsive axon guidance protein, draxin, FLJ34999, MGC117222, neucrin, UNQ3119
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Draxin/C1orf187 (NP_940947.3; Met1-Val349).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374946
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.