Learn More
Description
Specifications
Specifications
| Antigen | Draxin/C1orf187 |
| Applications | Western Blot, ELISA, Dot Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:20000-1:50000, Dot Blot 1:20000-1:50000, Immunocytochemistry 1:100 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | AGPA3119, C1orf187, chromosome 1 open reading frame 187, dorsal inhibitory axon guidance protein, Dorsal repulsive axon guidance protein, draxin, FLJ34999, MGC117222, neucrin, UNQ3119 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 300-330 on C terminal region of Human Draxin protein. Region: NNKCFDDCMC VEGLRCYAKF HRNRRVTRRK |
| Purification Method | Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
