Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DreamTaq DNA Polymerase (5 U/μL) PROMO
Get higher sensitivity, longer PCR products and higher yields in all standard PCR applications with this enhanced Taq DNA polymerase.
Supplier: Thermo Scientific™ EP0701
Description
Highlights
- Robust amplification with minimal optimization
- High yields of PCR products
- Higher sensitivity compared to conventional Taq DNA polymerase
- Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
- Incorporates modified nucleotides, but does not incorporate dUTP
Specifications
Contains: 40 μL DreamTaq DNA Polymerase (5 U/μL), 1.25 mL 10X DreamTaq Buffer (includes 20 mM MgCl2), Store at -20°C. | |
DreamTaq DNA Polymerase | |
DreamTaq™ | |
1X | |
3'-A |
Low | |
Standard | |
200 Units | |
No | |
Standalone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction