Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drebrin 1 Antibody (DBN-N-03), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP222344W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222344W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222344W Supplier Novus Biologicals Supplier No. NBP222344AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Drebrin 1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone DBN-N-03
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry
Gene Alias D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1
Gene Symbols DBN1
Host Species Mouse
Immunogen Bacterially expressed N-terminal fragment of recombinant human Drebrin Antibody (DBN-N-03)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1627
Test Specificity The mouse monoclonal Drebrin Antibody (DBN-N-03) recognizes drebrin, an approximately 100-125 kDa regulator of actin cytoskeleton.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.