Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drebrin 1 Antibody (DBN1/2879), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB012167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012167 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012167 Supplier Novus Biologicals Supplier No. NBP279901AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Drebrin 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DBN1/2879
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1
Gene Symbols DBN1
Host Species Mouse
Immunogen Recombinant fragment (aa 150-281) of human Drebrin 1 protein (exact sequence is proprietary) (Uniprot: Q16643)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.