Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drebrin 1, Mouse anti-Human, Clone: DBN1/2880, Novus Biologicals™
SDP

Catalog No. NB010552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010552 20 μg
NB010551 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010552 Supplier Novus Biologicals Supplier No. NBP27976720UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Drebrin 1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone DBN1/2880
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q16643
Gene Alias D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1
Gene Symbols DBN1
Host Species Mouse
Immunogen Recombinant fragment (aa 150-281) of human Drebrin 1 protein (exact sequence is proprietary)
Purification Method N/A
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1627
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.