Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DREF Antibody (OTI5A2), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271419B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271419B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271419B Supplier Novus Biologicals Supplier No. NBP271419B

Mouse Monoclonal Antibody

DREF Monoclonal specifically detects DREF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen DREF
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI5A2
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Ac-like transposable element, ALTEDNA replication-related element binding factor, hDREF, KIAA0785dREF homolog, Putative Ac-like transposable element, putative Ac-like transposon, TRAMPDREFBED-type zinc finger domain-containing protein 1, zinc finger BED domain-containing protein 1, zinc finger, BED-type containing 1
Gene Symbols ZBED1
Host Species Mouse
Immunogen Full length recombinant protein of human DREF (NP_004720) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9189.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.