Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DREF Antibody (OTI5A2), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB274979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274979 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274979 Supplier Novus Biologicals Supplier No. NBP271419MFV610

Mouse Monoclonal Antibody

DREF Monoclonal antibody specifically detects DREF in Human samples. It is validated for Flow Cytometry

Specifications

Antigen DREF
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5A2
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Ac-like transposable element, ALTEDNA replication-related element binding factor, hDREF, KIAA0785dREF homolog, Putative Ac-like transposable element, putative Ac-like transposon, TRAMPDREFBED-type zinc finger domain-containing protein 1, zinc finger BED domain-containing protein 1, zinc finger, BED-type containing 1
Host Species Mouse
Immunogen Full length human recombinant protein of human ZBED1(NP_004720) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9189
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.