Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRGX Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343997 Supplier Novus Biologicals Supplier No. NBP241138MFV610

Chicken Polyclonal Antibody

DRGX Polyclonal antibody specifically detects DRGX in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen DRGX
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias dorsal root ganglia homeobox, DRG11, paired related homeobox-like 1, paired-like homeodomain trancription factor DRG11, paired-related homeobox protein-like 1, PRRXL1
Host Species Chicken
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human DRGX. The immunogen is located within the last 50 amino acids of DRGX. Amino Acid Squence: VAKPAPPDGSQEKTSPTKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 644168
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.