Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell AlphaTec™ 1500 PLUS - Lab Coat Model 208
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
3X-Large
4X-Large
5X-Large
Description
- Short style, white lab coats
- SMS fabric construction
- Anti-static treatment
- Air and water vapor permeable
- Silicone-free, bound seams and low-linting
- Elastic cuffs, studded front-fastening and left-breast & right-thigh integral pockets
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Size | Medium |
| Gender | Unisex |
| Material | SMS |
| Color | White |
| Resistance | Static |
| Collar Type | Serged |
| Certifications/Compliance (Europe) | CE CATEGORY III (Type PB [6]) |
| For Use With (Application) | Unpacking Bulk Goods, Loading and Unloading Processing Equipment, Production Line Support and Maintenance, Assembly and Inspection of Components |
| No. per Pack | 70 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
