Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIL1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB057049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057049 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057049 Supplier Novus Biologicals Supplier No. NBP298981C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DRIL1 Polyclonal specifically detects DRIL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DRIL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARID domain-containing 3A, ARID domain-containing protein 3A, AT rich interactive domain 3A (BRIGHT- like), AT rich interactive domain 3A (BRIGHT- like) protein, AT rich interactive domain 3A (BRIGHT-like), AT-rich interactive domain-containing protein 3A, B-cell regulator of IgH transcription, Bright, dead ringer-like 1, Dead ringer-like protein 1, DRIL1dead ringer-like 1 (Drosophila), DRIL3, DRX, E2F-binding protein 1, E2FBP1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human DRIL1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1820
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.