Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Dropping Funnels with Rotaflow Stopcock
SCI_ED

Catalog No. p-7147185 Shop All Eisco Products
Click to view available options
Capacity (Metric):
50 mL
Stopper Number:
19/26

High quality borosilicate 3.3 glass funnel with screw cap Rotaflow stopcocks and interchangeable polypropylene stoppers

  • Ungraduated dropping funnel
  • Made from high quality borosilicate 3.3 glass
  • Cylindrical shape
  • Screw cap Rotaflow stopcock
  • Includes interchangeable, chemical resistant polypropylene stopper
  • Used for titrations and neutralizations
  • Funnel and stopper both autoclavable; Do not autoclave Rotaflow screw cap
  • Warranty and Services

    1 year limited manufacturer warranty

    Specifications

    Capacity (Metric) 50 mL
    Material Borosilicate Glass
    Height (Metric) 15.2 cm
    Autoclavable Autoclavable
    Color Clear
    For Use With (Application) Titration, neutralization
    Includes funnel, stopper
    Quantity 1
    Stopcock Rotaflow
    Stopper Material Polypropylene
    Stopper Number 19/26
    Product Type Cylindrical Dropping Funnel
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.