Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drosha Rabbit anti-Human, Clone: 4R0B7, Novus Biologicals™
SDP

Catalog No. NB166703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166703 100 μg
NB166704 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166703 Supplier Novus Biologicals Supplier No. NBP315804100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Drosha Monoclonal antibody specifically detects Drosha in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Drosha
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 4R0B7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias drosha, double-stranded RNA-specific endoribonuclease, drosha, ribonuclease type III, EC 3.1.26.3, Etohi2, nuclear RNase III Drosha, p241, Protein Drosha, putative protein p241 which interacts with transcription factor Sp1, putative ribonuclease III, RANSE3L, ribonuclease 3, Ribonuclease III, ribonuclease III, nuclear, RN3nuclear, RNase III
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Drosha (Q9NRR4).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 29102
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.