Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSCC1 Antibody (V76P1B2*B12), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20380A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20380A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20380A94 Supplier Novus Biologicals Supplier No. NBP250380AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

DSCC1 Monoclonal specifically detects DSCC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen DSCC1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V76P1B2*B12
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DCC1MGC5528, defective in sister chromatid cohesion 1 homolog (S. cerevisiae), Defective in sister chromatid cohesion protein 1 homolog, hDCC1, sister chromatid cohesion protein DCC1
Gene Symbols DSCC1
Host Species Mouse
Immunogen Peptide Sequence - KVYNSRRPIS (immunogen peptide is identical in sheep, bovine, canine, equine, chicken and zebrafish)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 79075
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.