Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSCC1 Antibody (V76P1B2*B12), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309506 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309506 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309506 Supplier Novus Biologicals Supplier No. NBP250380MFV450

Mouse Monoclonal Antibody

DSCC1 Monoclonal antibody specifically detects DSCC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen DSCC1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V76P1B2*B12
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias DCC1MGC5528, defective in sister chromatid cohesion 1 homolog (S. cerevisiae), Defective in sister chromatid cohesion protein 1 homolog, hDCC1, sister chromatid cohesion protein DCC1
Host Species Mouse
Immunogen Peptide Sequence - KVYNSRRPIS (immunogen peptide is identical in sheep, bovine, canine, equine, chicken and zebrafish)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 79075
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.