Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX), 50 mg PROMO

Catalog No. FERA52286PM
Click to view available options
:
Thermo Scientific™ DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX)

Catalog No. FERA52286PM

Supplier: Thermo Scientific™ A52286

Improve mass spectrometry detection of crosslinked proteins and interactions with DSPP and TBDSPP heterotrifunctional crosslinkers.

  • Tri-functional crosslinker: NHS ester (both ends), phosphonic acid (DSPP), or phosphonate ester (TBDSPP) in the spacer for enrichment
  • High-purity crystalline reagents for protein structure and interaction characterization
  • Membrane-permeable version (TBDSPP) for intracellular crosslinking
  • Enrichable using Fe-NTA IMAC or TiO2 MOAC

DSPP (disuccinimidyl phenyl phosphonic acid, PhoX) and TBDSPP (tert-butyl disuccinimidyl phenyl phosphonate, tBu-PhoX) are phospho-enrichable crosslinkers containing an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of a 7-atom spacer arm with a phosphonic acid group (DSPP) or phosphonate ester (TBDSPP) for enrichment. Both react efficiently with primary amine groups (-NH2) in pH 7–9 buffers to form stable amide bonds. The phosphonic acid or phosphonate ester groups can then be used to enrich crosslinked peptides using immobilized metal affinity chromatography (IMAC) or metal oxide affinity chromatography (MOAC).

Of the two enrichable MS crosslinkers, DSPP is more water soluble than TBDSPP and is better for in vitro crosslinking, simple purified proteins and protein complexes. TBDSPP is better for intracellular crosslinking due to increased membrane permeability. Although both crosslinkers can be enriched using traditional phosphopeptide methods, TBDSPP crosslinked peptides must first be incubated with TFA to remove the tert-butyl protection groups. The TBDSPP phosphonic acid group (after acid cleavage) is not a substrate for phosphatases, which provides opportunity for the removal endogenous phosphopeptides in samples prior to enrichment.

Note: For high resolution analysis of the crosslinked peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164940 or 164568). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES900 or ES904).

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Molecular Weight (g/mol) 440.25 g/mol
PEGylated No
Purity 90%
Spacer Arm Length 4.3 Å
Content And Storage Upon receipt store at -20°C protected from moisture.
Cell Permeability No
Labeling Method Chemical Labeling
Crosslinker Type Homotrifunctional
Reactive Moiety NHS Ester
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.