Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsRNA Antibody (1D3), PerCP, Novus Biologicals™
SDP

Catalog No. NB340486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340486 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340486 Supplier Novus Biologicals Supplier No. NBP320110PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen dsRNA
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1D3
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS
Gene Alias Double-stranded RNA
Host Species Mouse
Immunogen RDV-RNA-methylated bovine serum albumin complex.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.