Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsRNA Antibody (J5), Biotin, Novus Biologicals™
SDP

Catalog No. NB270874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270874 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270874 Supplier Novus Biologicals Supplier No. NBP311396B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot

Specifications

Antigen dsRNA
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot
Classification Monoclonal
Clone J5
Conjugate Biotin
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting
Formulation PBS
Gene Alias Double-stranded RNA
Host Species Mouse
Immunogen Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.