Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsRNA Mouse anti-Broad Species Reactivity, Clone: K1, Novus Biologicals™
SDP

Catalog No. NB121498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121498 200 μg
NB121499 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB121498 Supplier Novus Biologicals Supplier No. NBP311397200UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot, Affinity Purification

Specifications

Antigen dsRNA
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot, Affinity Purification
Classification Monoclonal
Clone K1
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting, Immunoaffinity Purification
Formulation Lyophilised from PBS
Gene Alias Double-stranded RNA
Host Species Mouse
Immunogen Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution The lyophilised sample should be reconstituted with 200 ul (500 ul for 500 ug size) sterile distilled water. The mAb will then be in PBS without any stabilisers or preservatives at a concentration of 1 mg/ml.
Target Species Multi-species
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.