Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format
Click to view available options
Quantity:
50 mg
10 x 2 mg
1 g
Description
Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.
- Reactive groups: NHS ester (both ends)
- Reactive towards: amino groups (primary amines)
- Amine reactive sulfo-NHS ester reacts rapidly with any primary amine-containing molecule
- Membrane-permeable, allowing for intracellular crosslinking
- High purity, crystalline reagent can be used to create high-purity conjugates
- Non-cleavable
- Water-insoluble (dissolve first in DMF or DMSO); compare to BS3 (Sulfo-DSS)
Specifications
Specifications
| Chemical Reactivity | Amine-Amine |
| Cleavable | No |
| Description | DSS |
| Molecular Weight (g/mol) | 368.35 |
| PEGylated | No |
| Spacer Arm Length | 11.4 Å |
| Content And Storage | Upon receipt store desiccated at 4°C. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | No-Weigh |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction