Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSS1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001334 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001334 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001334 Supplier Novus Biologicals Supplier No. NB1001334
Add to Cart
Add to Cart

Goat Polyclonal Antibody

DSS1 Polyclonal specifically detects DSS1 in Other samples. It is validated for Peptide ELISA.

Specifications

Antigen DSS1
Applications Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006295.
Gene Alias 26S proteasome complex subunit DSS1, Deleted in split hand/split foot protein 1, deleted in split-hand/split-foot 1, DSS1Split hand/foot malformation type 1 protein, ECD, SEM1, Shfdg1, SHSF1SHFD1, Split hand/foot deleted protein 1, split hand/foot malformation (ectrodactyly) type 1
Gene Symbols SEM1
Host Species Goat
Immunogen Peptide with sequence C-RAELEKHGYKMETS corresponding to C-Terminus according to NP_006295.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7979
Target Species Other
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.