Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dtk/TYRO3 Antibody (109646), FITC, Novus Biologicals™
SDP

Catalog No. NB302719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302719 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302719 Supplier Novus Biologicals Supplier No. FAB759F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Dtk/TYRO3 Monoclonal antibody specifically detects Dtk/TYRO3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Immunocytochemistry, CyTOF

Specifications

Antigen Dtk/TYRO3
Applications Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 109646
Conjugate FITC
Dilution Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Tyro3/Dtk, Ala31-Ser418, Accession # P55144
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 7301
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.