Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dtk/TYRO3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP12863A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP12863A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP12863A30 Supplier Novus Biologicals Supplier No. NBP128635AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dtk/TYRO3 Polyclonal specifically detects Dtk/TYRO3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).

Specifications

Antigen Dtk/TYRO3
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Gene Symbols TYRO3
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-ProtNo. Q06418]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7301
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.