Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dtk/TYRO3 Antibody (OTI2C4), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288961 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288961 Supplier Novus Biologicals Supplier No. NBP271331APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dtk/TYRO3 Monoclonal antibody specifically detects Dtk/TYRO3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Dtk/TYRO3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone OTI2C4
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Host Species Mouse
Immunogen Full length human recombinant protein of human TYRO3 (NP_006284) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 7301
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.