Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTX1/DTX4 Antibody (730612), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB375051 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375051 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375051 Supplier Novus Biologicals Supplier No. FAB7157W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DTX1/DTX4 Monoclonal antibody specifically detects DTX1/DTX4 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen DTX1/DTX4
Applications Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 730612
Conjugate DyLight 650
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DTX1;Fxit1;hDx-1;mKIAA4160
Host Species Mouse
Immunogen E. coli-derived recombinant human DTX1/DTX4, Met1-Phe147, Accession # Q86Y01
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.