Learn More
DTYMK Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305652AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Thymidylate kinase (DTYMK) catalyzes the phosphorylation of dTMP to form dTDP in the dTTP synthesis pathway for DNA synthesis. DTYMK is essential for DNA synthesis and is an important intermediate enzyme in the pathway of many pyrimidine analog drugs, including 5-Fluorouracil (5-FU), the chemotherapeutic drug of choice for several solid tumors. The human DTYMK gene was isolated via functional complementation of a Saccharomyces cerevisiae cell cycle mutant, cdc8. DTYMK mRNA levels and enzyme activities are regulated at the level of cell cycle and cell growth pathways.Specifications
DTYMK | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P23919, P97930 | |
DTYMK | |
Between 162 and 212 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
DTYMK | |
Thymidylate kinase; dTMP kinase; CDC8; TMPK; TYMK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1841, 21915 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.