Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUOX2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N1106176A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1106176A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1106176A94 Supplier Novus Biologicals Supplier No. NB11061576AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DUOX2 Polyclonal specifically detects DUOX2 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen DUOX2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias dual oxidase 2, dual oxidase-like domains 2, EC 1.11.1.-, EC 1.6.3.1, flavoprotein NADPH oxidase, Large NOX 2, LNOX2P138(TOX), Long NOX 2, NADH/NADPH thyroid oxidase p138-tox, NADPH oxidase/peroxidase DUOX2, NADPH thyroid oxidase 2, nicotinamide adenine dinucleotide phosphate oxidase, NOXEF2, p138 thyroid oxidase, P138-TOX, THOX2TDH6, Thyroid oxidase 2
Gene Symbols DUOX2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human DUOX2 (within residues 400-500). [Swiss-ProtNo. Q9NRD8]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 50506
Target Species Human, Mouse, Rat, Pig, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.