Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP14 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB322870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322870 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322870 Supplier Novus Biologicals Supplier No. NBP299994PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DUSP14 Polyclonal antibody specifically detects DUSP14 in Human samples. It is validated for ELISA

Specifications

Antigen DUSP14
Applications ELISA
Classification Polyclonal
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias dual specificity phosphatase 14, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 6, Mitogen-activated protein kinase phosphatase 6, MKP-1 like protein tyrosine phosphatase, MKP-1-like protein tyrosine phosphatase, MKP-6, MKP6MKP-Ldual specificity protein phosphatase 14
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human DUSP14 (Accession#: O95147; Met1-His191)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 11072
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.