Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP16 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224501SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224501SS 0.025 mg
NBP224501 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224501SS Supplier Novus Biologicals Supplier No. NBP224501SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DUSP16 Polyclonal specifically detects DUSP16 in Human, Mouse, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DUSP16
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Accession No. Q9BY84
Gene Alias dual specificity phosphatase 16, EC 3.1.3.16, EC 3.1.3.48, FLJ36298, FLJ40991, KIAA1700MGC129701, MAP kinase phosphatase 7, MAPK phosphatase-7, Mitogen-activated protein kinase phosphatase 7, MKP7dual specificity protein phosphatase 16, MKP-7MGC129702
Gene Symbols DUSP16
Host Species Rabbit
Immunogen A portion of amino acids 600-650 of human DUSP16 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80824
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), rhesus monkey and chimpanzee, 92% homologous in mouse, 84% homologous in rat and 76% homologous in dog.
Target Species Human, Mouse, Bovine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.