Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP16 Antibody, Novus Biologicals™
SDP

Catalog No. NB100848 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100848 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100848 Supplier Novus Biologicals Supplier No. NB100848
Add to Cart
Add to Cart

Goat Polyclonal Antibody

DUSP16 Polyclonal specifically detects DUSP16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA.

Specifications

Antigen DUSP16
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_085143.1
Gene Alias dual specificity phosphatase 16, EC 3.1.3.16, EC 3.1.3.48, FLJ36298, FLJ40991, KIAA1700MGC129701, MAP kinase phosphatase 7, MAPK phosphatase-7, Mitogen-activated protein kinase phosphatase 7, MKP7dual specificity protein phosphatase 16, MKP-7MGC129702
Gene Symbols DUSP16
Host Species Goat
Immunogen Peptide with sequence AHEMIGTQIVTER-C corresponding to N-Terminus according to NP_085143.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80824
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.