Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP22 Antibody, Novus Biologicals™
SDP

Catalog No. NBP183078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP183078 0.1 mL
NB433195 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP183078 Supplier Novus Biologicals Supplier No. NBP183078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DUSP22 Polyclonal specifically detects DUSP22 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DUSP22
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias dual specificity phosphatase 22, dual specificity protein phosphatase 22, EC 3.1.3.16, EC 3.1.3.48, FLJ35864, homolog of mouse dual specificity phosphatase LMW-DSP2, JKAP, JNK-stimulating phosphatase 1, JSP-1, JSP1JNK-stimulatory phosphatase-1, LMWDSP2, LMW-DSP2, Low molecular weight dual specificity phosphatase 2, Mitogen-activated protein kinase phosphatase x, MKP-x, MKPXMAP kinase phosphatase x, VHX
Gene Symbols DUSP22
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:GVSRSVTLVIAYIMTVTDFGWEDALHTVRAGRSCANPNVGFQRQLQEFEKHEVHQYRQWLKEEYGESPLQDAEEAKNILAAPGILKFW
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, MAP Kinase Signaling, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 56940
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.