Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP23 Antibody (OTI3C10), PE, Novus Biologicals™
SDP

Catalog No. NBP271976PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271976PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271976PE Supplier Novus Biologicals Supplier No. NBP271976PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DUSP23 Monoclonal specifically detects DUSP23 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen DUSP23
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3C10
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias dual specificity phosphatase 23, dual specificity protein phosphatase 23, DUSP25, EC 3.1.3.16, EC 3.1.3.48, FLJ20442, LDP3, LDP-3, Low molecular mass dual specificity phosphatase 3, low-molecular-mass dual-specificity phosphatase 3, MOSP, RP11-190A12.1, VH1-like member Z, VH1-like phosphatase Z, VHZ
Gene Symbols DUSP23
Host Species Mouse
Immunogen Full length recombinant protein of human DUSP23 (NP_060293) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline GPCR, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 54935.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.