Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP23 Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB045608 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045608 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045608 Supplier Novus Biologicals Supplier No. NBP297610AF405

Rabbit Polyclonal Antibody

DUSP23 Polyclonal specifically detects DUSP23 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DUSP23
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias dual specificity phosphatase 23, dual specificity protein phosphatase 23, DUSP25, EC 3.1.3.16, EC 3.1.3.48, FLJ20442, LDP3, LDP-3, Low molecular mass dual specificity phosphatase 3, low-molecular-mass dual-specificity phosphatase 3, MOSP, RP11-190A12.1, VH1-like member Z, VH1-like phosphatase Z, VHZ
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DUSP23 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 54935
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.