Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP26 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB181727 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181727 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181727 Supplier Novus Biologicals Supplier No. NBP306507AF750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DUSP26 Polyclonal antibody specifically detects DUSP26 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen DUSP26
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DSP-4, dual specificity phosphatase 26 (putative), Dual specificity phosphatase SKRP3, dual specificity protein phosphatase 26, dual-specificity phosphatase SKRP3, DUSP24SKRP3, EC 3.1.3.16, EC 3.1.3.48, LDP4, LDP-4, Low-molecular-mass dual-specificity phosphatase 4, MAP kinase phosphatase 8, MGC1136, MGC2627, Mitogen-activated protein kinase phosphatase 8, MKP8, MKP-8, NATA1, Novel amplified gene in thyroid anaplastic cancer
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DUSP26 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 78986
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.