Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP26 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB351615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351615 Supplier Novus Biologicals Supplier No. NBP306507MFV450

Rabbit Polyclonal Antibody

DUSP26 Polyclonal antibody specifically detects DUSP26 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen DUSP26
Applications Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DSP-4, dual specificity phosphatase 26 (putative), Dual specificity phosphatase SKRP3, dual specificity protein phosphatase 26, dual-specificity phosphatase SKRP3, DUSP24SKRP3, EC 3.1.3.16, EC 3.1.3.48, LDP4, LDP-4, Low-molecular-mass dual-specificity phosphatase 4, MAP kinase phosphatase 8, MGC1136, MGC2627, Mitogen-activated protein kinase phosphatase 8, MKP8, MKP-8, NATA1, Novel amplified gene in thyroid anaplastic cancer
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DUSP26 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 78986
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.