Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP27/DUPD1 Antibody (OTI7E4), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272503A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272503A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272503A405 Supplier Novus Biologicals Supplier No. NBP272503AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DUSP27/DUPD1 Monoclonal specifically detects DUSP27/DUPD1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen DUSP27/DUPD1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI7E4
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias atypical dual-specific protein phosphatase, Dual specificity phosphatase 27, dual specificity phosphatase and pro isomerase domain containing 1, DUSP27dual specificity phosphatase DUPD1, EC 3.1.3.16, EC 3.1.3.48, FMDSP
Gene Symbols DUPD1
Host Species Mouse
Immunogen Full length recombinant protein of human DUSP27/DUPD1 (NP_001003892) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 338599.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.