Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP4 Antibody (OTI7C11), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289488 Supplier Novus Biologicals Supplier No. NBP270610APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DUSP4 Monoclonal antibody specifically detects DUSP4 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen DUSP4
Applications Western Blot
Classification Monoclonal
Clone OTI7C11
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias dual specificity phosphatase 4, Dual specificity protein phosphatase hVH2, EC 3.1.3.16, EC 3.1.3.48, HVH2serine/threonine specific protein phosphatase, MAP kinase phosphatase 2, Mitogen-activated protein kinase phosphatase 2, MKP-2dual specificity protein phosphatase 4, MKP2VH1 homologous phosphatase 2, TYP, VH2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-257 of human DUSP4(NP_476499) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1846
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.