Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15830220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15830220 20 μL
NBP158302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15830220 Supplier Novus Biologicals Supplier No. NBP15830220UL

Rabbit Polyclonal Antibody

DUSP8 Polyclonal specifically detects DUSP8 in Human samples. It is validated for Western Blot.

Specifications

Antigen DUSP8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q13202
Gene Alias C11orf81, chromosome 11 open reading frame 81, dual specificity phosphatase 8, Dual specificity protein phosphatase hVH-5, EC 3.1.3.16, EC 3.1.3.48, FLJ42476, FLJ42958, HVH8, serine/threonine specific protein phosphatase, vaccinia virus homolog, VH5
Gene Symbols DUSP8
Host Species Rabbit
Immunogen Synthetic peptides corresponding to DUSP8(dual specificity phosphatase 8) The peptide sequence was selected from the middle region of DUSP8. Peptide sequence PAPPTPPATSALQQGLRGLHLSSDRLQDTNRLKRSFSLDIKSAYAPSRRP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1850
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.