Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUX4 Antibody (P2B1), FITC, Novus Biologicals™
SDP

Catalog No. NBP212886F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212886F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212886F Supplier Novus Biologicals Supplier No. NBP212886F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DUX4 Monoclonal specifically detects DUX4 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DUX4
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone P2B1
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10
Gene Symbols DUX4
Host Species Mouse
Immunogen C-terminal 76 amino acids of DUX4 with glutathione-s-transferase (gst) tag
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 100288687
Test Specificity Detects approx 45kDa. No cross-reactivity with DUX4c.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.