Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUX4 Antibody (P4H2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP149552SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149552SS 0.025 mL
NBP149552 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP149552SS Supplier Novus Biologicals Supplier No. NBP149552SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 5 publications

DUX4 Monoclonal specifically detects DUX4 in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen DUX4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone P4H2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2.0ug/ml, Simple Western 1:10, ELISA reported in scientific literature (PMID 21529284), Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200
Gene Accession No. Q9UBX2
Gene Alias double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10
Gene Symbols DUX4
Host Species Mouse
Immunogen Synthetic peptide corresponding to the C-terminus of human DUX4. [Swiss-Prot: Q9UBX2]
Molecular Weight of Antigen 52 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 100288687
Test Specificity This antibody is specific for DUX4 and does not recognize DUX4c.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.