Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUX4 Antibody (P4H2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$206.50 - $514.00
Specifications
Antigen | DUX4 |
---|---|
Clone | P4H2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5-2.0ug/ml, Simple Western 1:10, ELISA reported in scientific literature (PMID 21529284), Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP149552SS
![]() |
Novus Biologicals
NBP149552SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP149552
![]() |
Novus Biologicals
NBP149552 |
0.1 mL |
Each for $514.00
|
|
|||||
Description
DUX4 Monoclonal specifically detects DUX4 in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
DUX4 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Neuroscience, Transcription Factors and Regulators | |
Q9UBX2 | |
100288687 | |
Synthetic peptide corresponding to the C-terminus of human DUX4. [Swiss-Prot: Q9UBX2] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
52 kDa |
P4H2 | |
Western Blot 0.5-2.0ug/ml, Simple Western 1:10, ELISA reported in scientific literature (PMID 21529284), Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10 | |
DUX4 | |
IgG1 | |
Protein G purified | |
This antibody is specific for DUX4 and does not recognize DUX4c. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title