Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DVL3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7205084 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8749 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8749 Supplier Proteintech Group Inc Supplier No. 134441AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of a multi-gene family which shares strong similarity with the Drosophila disheveled gene, dsh. The Drosophila disheveled gene encodes a cytoplasmic phosphoprotein that regulates cell proliferation.

Specifications

Antigen DVL3
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DVL3
Gene Accession No. Q92997
Gene Alias Dishevelled 3, DSH homolog 3, DVL3, KIAA0208
Gene Symbols DVL3
Host Species Rabbit
Immunogen DVL3 Fusion Protein Ag4272
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1857
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.