Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DY 268
GREENER_CHOICE

Catalog No. p-7110862
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent FXR antagonist

DY 268 is a potent FXR antagonist (IC50 = 7.5 nM). Exhibits no detectable cytotoxicity in cell-based assays.

Specifications

Chemical Name or Material 1-[(3-Methoxyphenyl)methyl]-N-[4-methyl-3-(4-morpholinylsulfonyl)phenyl]-3-(4-methylphenyl)-1H-pyrazole-4-carboxamide
CAS 1609564-75-1
Quantity 10 mg
Target LXR-like Receptor Antagonists
Molecular Formula C30H32N4O5S
Formula Weight 560.66
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.