Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYDC2 Antibody (101), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB038290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038290 Supplier Novus Biologicals Supplier No. NBP290258AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DYDC2 Monoclonal specifically detects DYDC2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DYDC2
Applications Immunofluorescence
Classification Monoclonal
Clone 101
Conjugate Alexa Fluor 488
Dilution Immunocytochemistry/Immunofluorescence, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human DYDC2 (Q96IM9; Thr102-Pro176).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84332
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.